Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSTC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595894 Supplier Invitrogen™ Supplier No. PA595894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OSTC Polyclonal Antibody for Western Blot, ELISA

OSTC is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSTC
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene OSTC
Gene Accession No. B0K025, Q78XF5, Q9NRP0
Gene Alias 2310008M10Rik; 5730557H03Rik; DC2; HDCMD45P; HSPC307; Hydrophobic protein HSF-28; oligosaccharyltransferase complex non-catalytic subunit; oligosaccharyltransferase complex subunit; oligosaccharyltransferase complex subunit (non-catalytic); oligosaccharyltransferase complex subunit OSTC; Ostc; ostc {ECO:0000250; RGD1560708; UniProtKB:Q9NRP0}
Gene Symbols OSTC
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362040, 58505, 66357
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.