Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Osteoactivin/GPNMB Antibody (105), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289886AF750
Description
Osteoactivin/GPNMB Monoclonal antibody specifically detects Osteoactivin/GPNMB in Human samples. It is validated for ELISASpecifications
Osteoactivin/GPNMB | |
Monoclonal | |
Alexa Fluor 750 | |
glycoprotein (transmembrane) nmb, glycoprotein NMB, HGFINglycoprotein nmb-like protein, NMBosteoactivin, Transmembrane glycoprotein HGFIN, transmembrane glycoprotein NMB | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Osteoactivin/GPNMB (Uniprot#: Q14956-2; Met1-Pro474). | |
0.1 mL | |
Cell Cycle and Replication | |
10457 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
105 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction