Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Osteoadherin/OSAD/OMD Antibody (014), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289674IR
Description
Osteoadherin/OSAD/OMD Monoclonal antibody specifically detects Osteoadherin/OSAD/OMD in Human samples. It is validated for ELISASpecifications
Osteoadherin/OSAD/OMD | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
4958 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
014 | |
ELISA | |
Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2Costeoadherin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Osteoadherin/OSAD/OMD (NP_005005.1; Met1-Glu421). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction