Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Osteocalcin Antibody (2H9F11F8), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062801UV
Description
Osteocalcin Monoclonal specifically detects Osteocalcin in Human, Rat, Porcine, Bovine, Rabbit, Mouse (Negative) samples. It is validated for ELISA, Immunohistochemistry.Specifications
Osteocalcin | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BGLAP | |
Recombinant human osteocalcin protein | |
0.1 mL | |
Extracellular Matrix, Stem Cell Markers | |
632 | |
Human, Rat, Pig, Bovine, Rabbit, Mouse (Negative) | |
Purified |
ELISA, Immunohistochemistry | |
2H9F11F8 | |
ELISA, Immunohistochemistry | |
BGP, bone gamma-carboxyglutamate (gla) protein, bone gamma-carboxyglutamate (gla) protein (osteocalcin), Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, OC, OCN, osteocalcin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
This product recognizes intact human and bovine osteocalcin in a “2 site” assay and reacts with osteocalcin fragments corresponding to amino acids 15-31 and 20-43 of the native molecule. It does not recognize fragments representing amino acids 1-19 or 7-19. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction