Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteocalcin (Glu17) Mouse anti-Human, Unconjugated, Clone: 4B6, MilliporeSigma™

Catalog No. MABS19672MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS19672MI 25 μL
MABS19671MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS19672MI Supplier MilliporeSigma Supplier No. MABS196725UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Osteocalcin (Glu17) Monoclonal antibody specifically detects Osteocalcin (Glu17) in Human samples. It is validated for ELISA, Dot Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osteocalcin (Glu17)
Applications ELISA, Dot Blot
Classification Monoclonal
Clone 4B6
Conjugate Unconjugated
Gene Accession No. NP_954642.1
Gene Alias Bone Gla protein;BGP;Gamma-carboxyglutamic acid-containing protein
Gene Symbols BGLAP
Host Species Mouse
Immunogen A linear peptide corresponding to 17 amino acids from the central region of the mature human osteocalcin with decarboxylated GLU17 residue (with signal peptide and pro regions cleaved off).
Molecular Weight of Antigen 10.96 kDa calculated (for complete molecule)
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 4B6 is a mouse monoclonal antibody that detects human osteocalcin only when not carboxylated on the first glutamic acid residue (GLU17). It does not react with carboxylated osteocalcin.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.