Learn More
Invitrogen™ Osteocalcin Monoclonal Antibody (5021)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA143028
Description
MA1-43028 detects Osteocalcin in human and bovine samples. MA1-43028 has successfully been used in radioimmunoassays, western blot, ELISA and immunohistochemistry applications. Immunohistochemistry is calcium dependent. MA1-43028 was derived from human bone osteocalcin.
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
Specifications
Osteocalcin | |
Monoclonal | |
5.7 mg/mL | |
50% water with 50% glycerol and no preservative | |
P02818, P02820 | |
BGLAP | |
Bone osteocalcin. | |
100 μg | |
Primary | |
Human, Bovine | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot | |
5021 | |
Unconjugated | |
BGLAP | |
Bglap; Bglap1; Bglap2; BGP; Bgpr; Bgpra; bone gamma carboxyglutamate protein; bone gamma carboxyglutamate protein 1; bone gamma-carboxyglutamate (gla) protein; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein; bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamate protein osteocalcin; bone gamma-carboxyglutamate; BGLAP; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); bone Gla protein; bone gla protein BGP; bone Gla-protein; gamma-carbooxyglutamate (gla) protein; Gamma-carboxyglutamic acid-containing protein; GLA; Gla protein precusor; mOC-A; OC; OCN; OG1; Osteocalcin; PMF1; RP11-54H19.5; unnamed protein product | |
Mouse | |
SDS-PAGE | |
RUO | |
281646, 632 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.