Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteocalcin Monoclonal Antibody (OC4-30), QED Bioscience™
SDP

Catalog No. 502278114
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8114 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8114 Supplier Leinco Technologies Inc Supplier No. 50114100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Osteocalcin Monoclonal antibody specifically detects Osteocalcin in Canine, Chicken, Goat, Human, Ovine, Porcine, Rabbit, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Osteocalcin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OC4-30
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and no preservative; pH 7.4
Gene BGLAP
Gene Accession No. P02818, P02822, P04640, P0C225, P39056, P81455, Q8HYY9
Gene Alias Bglap; Bglap1; Bglap2; BGP; Bgpr; Bgpra; bone gamma carboxyglutamate protein; bone gamma carboxyglutamate protein 1; bone gamma-carboxyglutamate (gla) protein; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein; bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamate protein osteocalcin; bone gamma-carboxyglutamate; BGLAP; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); bone Gla protein; bone gla protein BGP; bone Gla-protein; gamma-carbooxyglutamate (gla) protein; Gamma-carboxyglutamic acid-containing protein; GLA; Gla protein precusor; mOC-A; OC; OCN; OG1; Osteocalcin; PMF1; RP11-54H19.5; unnamed protein product
Host Species Mouse
Immunogen Bovine osteocalcin
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100340503, 100861188, 25295, 396348, 397530, 403762, 632, 677670
Target Species Canine, Chicken, Goat, Human, Ovine, Pig, Rabbit, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.