Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Osteocrin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121446
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
Hormone that acts as a regulator of dendritic growth in the developing cerebral cortex in response to sensory experience. Induced in the brain following membrane depolarization and inhibits dendritic branching in neurons of the developing cortex.
Specifications
Osteocrin | |
Polyclonal | |
Unconjugated | |
OSTN | |
MUSCLIN; Ostc; osteocrin; Ostn; Processed Osteocrin | |
Rabbit | |
Protein G | |
RUO | |
344901 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
2.63 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P61366 | |
OSTN | |
Recombinant Human Osteocrin protein (28-133aa). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction