Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteopontin Mouse anti-Human,Mouse, Unconjugated, Clone: 2A1, MilliporeSigma™

Catalog No. MABT15171MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT15171MI 100 μg
MABT15172MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT15171MI Supplier MilliporeSigma Supplier No. MABT1517100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Osteopontin Monoclonal antibody specifically detects Osteopontin in Human,Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoaffinity Purification, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osteopontin
Applications ELISA, Immunohistochemistry, Affinity Purification, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2A1
Conjugate Unconjugated
Gene Accession No. NP_000573
Gene Alias Bone sialoprotein 1;Nephropontin;Secreted phosphoprotein 1;SPP-1;Urinary stone protein;OPN;Uropontin
Gene Symbols SPP1, BNSP, OPN, PSEC0156
Host Species Mouse
Immunogen His-tagged full-length recombinant human Osteopontin.
Molecular Weight of Antigen ∼26 kDa observed; 35.42 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone 2A1 is a mouse monoclonal antibody that specifically detects Osteopontin. It targets an epitope within the C-terminal half.
Target Species Human, Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.