Learn More
Invitrogen™ Osteopontin Polyclonal Antibody

Description
By Western Blot, a doublet band of approximately 65 kDa and a band of 30 kDa is observed due to glycosylation. A suggested positive control is 293 cells. The Neutralization Dose50 (ND50) for anti-mouse osteopontin is approximately 1 to 3 μg/mL in the presence of 2 μg/mL of recombinant mouse osteopontin using 293 cells.
Specifications
Specifications
| Antigen | Osteopontin |
| Applications | ELISA, Inhibition Assays, Immunohistochemistry, Neutralization, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and no preservative |
| Gene | SPP1 |
| Gene Accession No. | P10923 |
| Gene Alias | 2AR; 2b7; 44 kDa bone phosphoprotein; Apl-1; BNSP; Bone sialoprotein 1; Bone sialoprotein 1 (BSPI/BSP1); Bsp; BSPI; Calcium oxalate crystal growth inhibitor protein; CALPHA1 fusion; Early T lymphocyte activation 1 (ETA1); early T-lymphocyte activation 1; early T-lymphocyte activation 1 protein; Eta; ETA-1; HGNC:11255; immunoglobulin alpha 1 heavy chain constant region fusion protein; MGC110940; Minopontin; nephropontin; Op; Opn; Opnl; OSP; osteopontin; osteopontin/immunoglobulin alpha 1 heavy chain constant region fusion protein; osteopontin-like protein; PSEC0156; Ric; Secreted phosphoprotein 1; secreted phosphoprotein 1 (osteopontin bone sialoprotein I early T lymphocyte activation 1); secreted phosphoprotein 1 (osteopontin, bone sialoprotein I, early T-lymphocyte activation 1); Secreted phosphoprotein 1 (SPP1); secreted phosphoprotein 1 variant 6; Sialoprotein (osteopontin); SPP1; SPP-1; SPP1/CALPHA1 fusion; unnamed protein product; urinar; urinary stone protein; Uropontin |
| Gene Symbols | SPP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.