Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteopontin Polyclonal Antibody, FITC, Bioss
SDP

Catalog No. 502279977
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9977 Supplier Bioss Supplier No. BS23258RFITC
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Osteopontin Polyclonal antibody specifically detects Osteopontin in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)

Specifications

Antigen Osteopontin
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene SPP1
Gene Accession No. P08721, P10451
Gene Alias 2AR; 2b7; 44 kDa bone phosphoprotein; Apl-1; BNSP; Bone sialoprotein 1; Bone sialoprotein 1 (BSPI/BSP1); Bsp; BSPI; Calcium oxalate crystal growth inhibitor protein; CALPHA1 fusion; Early T lymphocyte activation 1 (ETA1); early T-lymphocyte activation 1; early T-lymphocyte activation 1 protein; Eta; ETA-1; HGNC:11255; immunoglobulin alpha 1 heavy chain constant region fusion protein; MGC110940; Minopontin; nephropontin; Op; Opn; Opnl; OSP; osteopontin; osteopontin/immunoglobulin alpha 1 heavy chain constant region fusion protein; osteopontin-like protein; PSEC0156; Ric; Secreted phosphoprotein 1; secreted phosphoprotein 1 (osteopontin bone sialoprotein I early T lymphocyte activation 1); secreted phosphoprotein 1 (osteopontin, bone sialoprotein I, early T-lymphocyte activation 1); Secreted phosphoprotein 1 (SPP1); secreted phosphoprotein 1 variant 6; Sialoprotein (osteopontin); SPP1; SPP-1; SPP1/CALPHA1 fusion; unnamed protein product; urinar; urinary stone protein; Uropontin
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from rat osteopontin (amino acids 201-314).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25353, 6696
Target Species Human, Rat
Content And Storage -20° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.