Learn More
Osteopontin Rabbit Polyclonal Antibody, Epredia™
Take advantage of a quality product that delivers accurate results with the Epredia™ Osteopontin, Rabbit Polyclonal Antibody.
Specifications
Antigen | Osteopontin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RB9097P0
|
Epredia
RB9097P0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
RB9097P1
|
Epredia
RB9097P1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
RB9097P
|
Epredia
RB9097P |
Concentrated | 1 mL | N/A | N/A | N/A | |||
RB9097R1
|
Epredia
RB9097R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
RB9097R7
|
Epredia
RB9097R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
Osteopontin is a 34kDa extracellular matrix protein with a cell-binding domain. It was originally identified as a major component of the non-collagenous organic bone matrix however it has subsequently been demonstrated in a wide range of normal adult tissue and body fluids. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. This antibody should prove useful in studying the expression of this matrix glycoprotein in tissue homeostasis. This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Pig and Dog. Others not tested.
Epitope: Internal domain
Immunogen: A synthetic peptide derived from internal domain of human Osteopontin
Molecular Weight: 34kDa
Positive Control: Jurkat cells; Gastric CA or Osteosarcoma
Cellular Localization: Cytoplasmic This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Osteopontin | |
Polyclonal | |
Rabbit | |
Extracellular Matrix | |
A synthetic peptide derived from internal domain of human Osteopontin | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.