Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osteoprotegerin (OPG) Mouse anti-Human, Clone: 98A1071, Active Motif™
SDP

Catalog No. 501993053
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3053 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-3053 Supplier Active Motif Supplier No. 40938
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.

Specifications

Antigen Osteoprotegerin (OPG)
Applications Western Blot
Classification Monoclonal
Clone 98A1071
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% gelatin and 0.05% sodium azide; pH 7.5
Gene TNFRSF11B
Gene Accession No. O00300
Gene Alias MGC29565; Ocif; OPG; Osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11b; TNFRSF11B; TR1; Tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b; tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin)
Gene Symbols TNFRSF11B
Host Species Mouse
Immunogen This Osteoprotegerin (OPG) antibody was raised against a peptide corresponding to amino acid residues 20-37 of human Osteoprotegerin (OPG).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4982
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.