Learn More
Oster™ Animal Clippers
Click to view available options
:
Blade size 10
Description
Includes
Size 10 blade set for general clipping
- Cutting blades snap on and off, permitting instant change with no tools required
- Universal motor uses high rpm and gear-down design to produce outstanding power for unit's size
- Automatic armature take-up ensures long life and quiet operation
For surgical shaving, use 01-305-10B Size 40 Blade Set (below).
Compliance
Double insulated to meet OSHA specifications.
Specifications
Specifications
Product Type | Single Speed Clipper |
For Use With (Equipment) | A5™ cryogen-x™ blade, elite cryogen-x™ blade and the wider take-down-quick™ wide blade series |
For Use With (Application) | Preparing mice, rats, hamsters or other animals for surgery without irritating or frightening them |
No. per Pack | 1 Ea. |