Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSTF1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150036AF647
Description
OSTF1 Polyclonal antibody specifically detects OSTF1 in Human samples. It is validated for Western BlotSpecifications
OSTF1 | |
Polyclonal | |
Western Blot | |
FLJ20559, OSFSH3P2bA235O14.1, osteoclast stimulating factor 1, osteoclast-stimulating factor 1 | |
The immunogen for this product maps to a region between residue 50 and 100 of human Osteoclast Stimulating Factor 1 using the numbering given in entry NP_036515.4 (GeneID 26578). | |
0.1 mL | |
Signal Transduction | |
26578 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction