Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSTF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588394 Supplier Invitrogen™ Supplier No. PA588394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVFRALYTFE PRTPDELYFE EGDIIYITDM SDTNWWKGTS KGRTGLIPSN YVAEQAESID NPLHEAAKRG NLSWLRECLD NRVGVNGLDK AGSTALYWAC HGGHKDIVEM LFTQPNIELN QQNKLGDTAL HAAAWKGYAD IVQLLLAKGA RTDLRNIEKK LAFDMATNAA CASLLKKKQG TDAVRTLSNA EDYLDDEDSD; Positive Samples: 293T, B-cell, HeLa, HepG2, Mouse brain, Rat kidney; Cellular Location: Cytoplasm.

Osteoclast-stimulating factor-1 is an intracellular protein produced by osteoclasts that indirectly induces osteoclast formation and bone resorption.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSTF1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ostf1
Gene Accession No. Q62422, Q6P686, Q92882
Gene Alias bA235O14.1; C78236; OSF; osteoclast stimulating factor 1; osteoclast-stimulating factor 1; OSTF1; SH3 domain protein 3; Sh3d3; SH3P2
Gene Symbols Ostf1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 15-214 of human OSTF1 (NP_0365154).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20409, 259275, 26578
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.