Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSTM1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299663MFV450
Description
OSTM1 Polyclonal antibody specifically detects OSTM1 in Human samples. It is validated for ELISASpecifications
OSTM1 | |
Polyclonal | |
ELISA | |
GIPN, GLGAIP-interacting protein N terminus, grey-lethal osteopetrosis, HSPC019, OPTB5, osteopetrosis associated transmembrane protein 1, osteopetrosis-associated transmembrane protein 1 | |
Produced in rabbits immunized with purified, recombinant Human OSTM1 (Accession#: NP_054747.2; Met1-Pro284) | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
28962 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction