Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSTM1 Monoclonal Antibody (2F2H10), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | OSTM1 |
---|---|
Clone | 2F2H10 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543502
|
Invitrogen™
MA543502 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543011
|
Thermo Scientific
MA543011 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human OSTM1
This gene encodes a protein that may be involved in the degradation of G proteins via the ubiquitin-dependent proteasome pathway. The encoded protein binds to members of subfamily A of the regulator of the G-protein signaling (RGS) family through an N-terminal leucine-rich region. This protein also has a central RING finger-like domain and E3 ubiquitin ligase activity. This protein is highly conserved from flies to humans. Defects in this gene may cause the autosomal recessive, infantile malignant form of osteopetrosis.Specifications
OSTM1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
1200002H13Rik; Chloride channel 7 beta subunit; CLCN7 accessory beta subunit; GAIP interacting protein N terminus; GAIP-interacting protein N terminus; GIPN; GL; grey lethal osteopetrosis; grey-lethal osteopetrosis; Grey-lethal protein; HSPC019; OPTB5; osteopetrosis associated transmembrane protein 1; osteopetrosis associated transmembrane protein 1-like; osteopetrosis-associated transmembrane protein 1; Ostm1; UNQ6098/PRO21201 | |
OSTM1 | |
IgG1 | |
Protein A | |
Antibody |
2F2H10 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q86WC4 | |
28962 | |
Recombinant Human OSTM1 protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
OSTM1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title