Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSTM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529176 Supplier Invitrogen™ Supplier No. PA529176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, HL-60. Predicted reactivity: Mouse (81%), Rat (83%), Rabbit (92%), Rhesus Monkey (99%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that may be involved in the degradation of G proteins via the ubiquitin-dependent proteasome pathway. The encoded protein binds to members of subfamily A of the regulator of the G-protein signaling (RGS) family through an N-terminal leucine-rich region. This protein also has a central RING finger-like domain and E3 ubiquitin ligase activity. This protein is highly conserved from flies to humans. Defects in this gene may cause the autosomal recessive, infantile malignant form of osteopetrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSTM1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene OSTM1
Gene Accession No. Q86WC4
Gene Alias 1200002H13Rik; Chloride channel 7 beta subunit; CLCN7 accessory beta subunit; GAIP interacting protein N terminus; GAIP-interacting protein N terminus; GIPN; GL; grey lethal osteopetrosis; grey-lethal osteopetrosis; Grey-lethal protein; HSPC019; OPTB5; osteopetrosis associated transmembrane protein 1; osteopetrosis associated transmembrane protein 1-like; osteopetrosis-associated transmembrane protein 1; Ostm1; UNQ6098/PRO21201
Gene Symbols OSTM1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human OSTM1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.