Learn More
Invitrogen™ OSTM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120130
Description
Positive test controls include: HeLa, 293T, A-549, Raji, B cells, Mouse brain, Mouse kidney. The target is usually found in the following locations: Lysosome membrane, Single-pass type I membrane protein. Immunogen sequence: ALPFGSSPHR VFHDLLSEQQ LLEVEDLSLS LLQGGGLGPL SLPPDLPDLD PECRELLLDF ANSSAELTGC LVRSARPVRL CQTCYPLFQQ VVSKMDNISR AAGNTSESQS CARSLLMADR MQIVVILSEF FNTTWQEANC ANCLTNNSEE LSNSTVYFLN LFNHTLTCFE HNLQGNAHSL LQTKNYSEVC KNCREAYKTL SSLYSEMQKM NELENKAEPG THLCIDVEDA MNITRKLWSR TFNCSVPCSD TVP.
This gene encodes a protein that may be involved in the degradation of G proteins via the ubiquitin-dependent proteasome pathway. The encoded protein binds to members of subfamily A of the regulator of the G-protein signaling (RGS) family through an N-terminal leucine-rich region. This protein also has a central RING finger-like domain and E3 ubiquitin ligase activity. This protein is highly conserved from flies to humans. Defects in this gene may cause the autosomal recessive, infantile malignant form of osteopetrosis.
Specifications
| OSTM1 | |
| Polyclonal | |
| Unconjugated | |
| OSTM1 | |
| 1200002H13Rik; Chloride channel 7 beta subunit; CLCN7 accessory beta subunit; GAIP interacting protein N terminus; GAIP-interacting protein N terminus; GIPN; GL; grey lethal osteopetrosis; grey-lethal osteopetrosis; Grey-lethal protein; HSPC019; OPTB5; osteopetrosis associated transmembrane protein 1; osteopetrosis associated transmembrane protein 1-like; osteopetrosis-associated transmembrane protein 1; Ostm1; UNQ6098/PRO21201 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14628, 28962 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.79 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q86WC4, Q8BGT0 | |
| OSTM1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 32-284 of human OSTM1 (NP_054747.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.