Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSTM1 Recombinant Rabbit Monoclonal Antibody (30)
GREENER_CHOICE

Catalog No. PIMA530728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530728 Supplier Invitrogen™ Supplier No. MA530728
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human OSTM1.

This gene encodes a protein that may be involved in the degradation of G proteins via the ubiquitin-dependent proteasome pathway. The encoded protein binds to members of subfamily A of the regulator of the G-protein signaling (RGS) family through an N-terminal leucine-rich region. This protein also has a central RING finger-like domain and E3 ubiquitin ligase activity. This protein is highly conserved from flies to humans. Defects in this gene may cause the autosomal recessive, infantile malignant form of osteopetrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSTM1
Applications ELISA
Classification Recombinant Monoclonal
Clone 30
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene OSTM1
Gene Accession No. Q86WC4
Gene Alias 1200002H13Rik; Chloride channel 7 beta subunit; CLCN7 accessory beta subunit; GAIP interacting protein N terminus; GAIP-interacting protein N terminus; GIPN; GL; grey lethal osteopetrosis; grey-lethal osteopetrosis; Grey-lethal protein; HSPC019; OPTB5; osteopetrosis associated transmembrane protein 1; osteopetrosis associated transmembrane protein 1-like; osteopetrosis-associated transmembrane protein 1; Ostm1; UNQ6098/PRO21201
Gene Symbols OSTM1
Host Species Rabbit
Immunogen Recombinant Human OSTM1 protein (Met1-Pro284).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.