Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OSU 03012
GREENER_CHOICE

Catalog No. 568210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-821-0 10 mg
56-825-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 56-821-0 Supplier Tocris Bioscience™ Supplier No. 5682/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

PDPK1 (PDK1) inhibitor; inhibits Akt signaling

OSU 03012 is a PDPK1 (PDK1) inhibitor; inhibits Akt signaling. Induces apoptosis of PC-3 and medulloblastoma cells, and inhibits growth of a number of tumor cell lines. Sensitizes radiotherapy-induced cell death and enhances cytotoxic effects of chemotherapeutic agents in vitro. Attenuates tumor growth of medulloblastoma xenografts in mice.

Specifications

Chemical Name or Material 2-Amino-N-[4-[5-(2-phenanthrenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]phenyl]acetamide
CAS 742112-33-0
Quantity 10 mg
Target PDK-1 Inhibitors
Molecular Formula C26H19F3N4O
Formula Weight 460.45
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.