Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OSU 6162 hydrochloride

Catalog No. 259910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-991-0 10 mg
25-995-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-991-0 Supplier Tocris Bioscience™ Supplier No. 2599/10
This item is not returnable. View return policy

Dopamine stabilizer

Dopamine stabilizer; lacks high affinity for various neuroreceptors in vitro (Ki values are 447 and 1305 nM for D2 and D3 receptors respectively and > 1 μM for other targets). Exhibits high D2 receptor occupancy in vivo; highly active on dopamine synthesis and turnover. Induces stabilizing effects on psychomotor function in behavioral tests without inducing hypolocomotion or catalepsy.

Chemical Identifiers

CAS 156907-84-5
Molecular Formula C15H24ClNO2S
Molecular Weight (g/mol) 317.872
InChI Key LEMGVHZVBREXAD-PFEQFJNWSA-N
PubChem CID 9836644
IUPAC Name (3S)-3-(3-methylsulfonylphenyl)-1-propylpiperidine;hydrochloride
SMILES CCCN1CCCC(C1)C2=CC(=CC=C2)S(=O)(=O)C.Cl

Specifications

Quantity 10 mg
Synonym osu 6162 hydrochloride, osu6162 hydrochloride, 3s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, pnu-96391; pnu-0096391; osu-6162; s---3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride; s,s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, 3s-3-3-methanesulfonylphenyl-1-propylpiperidine hydrochloride, unii-v5sbjbujw6, v5sbjbujw6, osu-6162 hydrochloride, osu6162 hydrochloride hplc, 3-3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride
Formula Weight 317.87
Percent Purity >98%
Chemical Name or Material OSU 6162 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.