Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTC Monoclonal Antibody (CL4045)
GREENER_CHOICE

Catalog No. PIMA531396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531396 Supplier Invitrogen™ Supplier No. MA531396
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: ILADYLTLQE HYSSLKGLTL SWIGDGNNIL HSIMMSAAKF GMHLQAATPK GYEPDASVTK LAEQYAKENG TKLLLTNDPL EAAHGGNVLI TDTWISMGQE EEKKKRLQAF QGYQVTMKTA KVAASDWT Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 92%.

This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL4045
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTC
Gene Accession No. P00480
Gene Alias AI265390; OCTD; ornithine carbamoyltransferase; ornithine carbamoyltransferase, mitochondrial; Ornithine transcarbamylase; Otc; OTCase; Sf; sparse fur; spf
Gene Symbols OTC
Host Species Mouse
Immunogen Recombinant protein fragment of Human OTC (Product #RP-91882)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5009
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.