Learn More
Invitrogen™ OTC Polyclonal Antibody

Description
Immunogen sequence: ILADYLTLQE HYSSLKGLTL SWIGDGNNIL HSIMMSAAKF GMHLQAATPK GYEPDASVTK LAEQYAKENG TKLLLTNDPL EAAHGGNVLI TDTWISMGQE EEKKKRLQAF QGYQVTMKTA KVAASDWT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
Specifications
Specifications
| Antigen | OTC |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | OTC |
| Gene Accession No. | P00480 |
| Gene Alias | AI265390; OCTD; ornithine carbamoyltransferase; ornithine carbamoyltransferase, mitochondrial; Ornithine transcarbamylase; Otc; OTCase; Sf; sparse fur; spf |
| Gene Symbols | OTC |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.