Learn More
Invitrogen™ OTC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595437
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat liver tissue, mouse liver tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.
Specifications
OTC | |
Polyclonal | |
Unconjugated | |
OTC | |
AI265390; OCTD; ornithine carbamoyltransferase; ornithine carbamoyltransferase, mitochondrial; Ornithine transcarbamylase; Otc; OTCase; Sf; sparse fur; spf | |
Rabbit | |
Affinity chromatography | |
RUO | |
18416, 25611, 5009 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P00480, P00481, P11725 | |
OTC | |
A synthetic peptide corresponding to a sequence at the N-terminus of human OTC (33-70aa NKVQLKGRDLLTLKNFTGEEIKYMLWLSADLKFRIKQK). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.