Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTOF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552935 Supplier Invitrogen™ Supplier No. PA552935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PIIVIEIYDQ DSMGKADFMG RTFAKPLVKM ADEAYCPPRF PPQLEYYQIY RGNATAGDLL AAFELLQIGP AGKADLPPIN GPVDVDRGPI MPVPMGIRPV LSKYRVEVLF WGLRDLKRVN L Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

Otoferlin is a protein that in humans is encoded by the OTOF gene. Mutations in this gene are a cause of neurosensory nonsyndromic recessive deafness, DFNB9. The short form of the encoded protein has three C2 domains, a single carboxy-terminal transmembrane domain found also in the C. elegans spermatogenesis factor FER-1 and human dysferlin, while the long form has six C2 domains. The homology suggests that this protein may be involved in vesicle membrane fusion. Several transcript variants encoding multipleisoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTOF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTOF
Gene Accession No. Q9HC10
Gene Alias AUNB1; DFNB6; DFNB9; FER1L2; fer-1-like family member 2; fer-1-like protein 2; NSRD9; Otof; Otoferlin
Gene Symbols OTOF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OTOF. Recombinant protein control fragment (Product #RP-90139).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9381
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.