Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Otoferlin Monoclonal Antibody (13A9), QED Bioscience™
SDP

Catalog No. 502278298
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8298 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8298 Supplier Leinco Technologies Inc Supplier No. 90023100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Otoferlin Monoclonal antibody specifically detects Otoferlin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Otoferlin
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 13A9
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris with 0.1M glycine, 2% sucrose and no preservative
Gene OTOF
Gene Accession No. Q9HC10
Gene Alias AUNB1; DFNB6; DFNB9; FER1L2; fer-1-like family member 2; fer-1-like protein 2; NSRD9; Otof; Otoferlin
Host Species Mouse
Immunogen GST fusion protein corresponding to aa 1-395 of human otoferlin (accession no. Q9HC10).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9381
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.