Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Otoferlin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579776
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579776 Supplier Invitrogen™ Supplier No. PA579776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Cardiac Muscle Tissue, 293T whole cell. IHC: mouse brain tissue, rat brain tissue, human glioma tissue.

Otoferlin is a protein that in humans is encoded by the OTOF gene. Mutations in this gene are a cause of neurosensory nonsyndromic recessive deafness, DFNB9. The short form of the encoded protein has three C2 domains, a single carboxy-terminal transmembrane domain found also in the C. elegans spermatogenesis factor FER-1 and human dysferlin, while the long form has six C2 domains. The homology suggests that this protein may be involved in vesicle membrane fusion. Several transcript variants encoding multipleisoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Otoferlin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation Antibody with 0.2mg Na2PO4, 0.9mg NaCl, 4mg trehalose and no preservative
Gene OTOF
Gene Accession No. Q9ERC5, Q9ESF1, Q9HC10
Gene Alias AUNB1; DFNB6; DFNB9; FER1L2; fer-1-like family member 2; fer-1-like protein 2; NSRD9; Otof; Otoferlin
Gene Symbols OTOF
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human Otoferlin (1831-1863aa QIWDADHFSADDFLGAIELDLNRFPRGAKTAKQ).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83762, 84573, 9381
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.