Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OTOG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144338
Description
OTOG Polyclonal Antibody for ICC/IF, IHC (P), ELISA
Glycoprotein specific to acellular membranes of the inner ear. May be required for the anchoring of the otoconial membranes and cupulae to the underlying neuroepithelia in the vestibule. May be involved in the organization and/or stabilization of the fibrillar network that compose the tectorial membrane in the cochlea. May play a role in mechanotransduction processes (By similarity).
Specifications
OTOG | |
Polyclonal | |
Unconjugated | |
OTOG | |
OTGN | |
Rabbit | |
Protein G | |
RUO | |
340990 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q6ZRI0 | |
OTOG | |
Recombinant Human Otogelin protein (1634-1796AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction