Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTOGL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571232 Supplier Invitrogen™ Supplier No. PA571232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 77%; Rabbit: 86%; Rat: 93%.

OTOGL is a protein coding gene. Gene Ontology (GO) annotations related to this gene include extracellular region; alpha-L-arabinofuranosidase activity; L-arabinose metabolic process; sensory perception of sound.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTOGL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OTOGL
Gene Accession No. Q3ZCN5
Gene Alias C12orf64; DFNB84B; EG628870; Gm6924; otogelin like; otogelin-like; otogelin-like protein; otogelin-like, pseudogene 1; Otogl; Otogl-ps1; RGD1564294
Gene Symbols OTOGL
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human OTOGL.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283310
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.