Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTOGL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565783 Supplier Invitrogen™ Supplier No. PA565783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CEVGEFTAIDH NFQSDCGCIQ YLCEKDDVCV FQEVSVLNPG QSMIKYLEED FCYTIECLEE KDNHTGFHTL NFTLVNCSKK CDVHQVYTPS PSDY Highest antigen sequence identity to the following orthologs - mouse 81%, rat 82%.

OTOGL is a protein coding gene. Gene Ontology (GO) annotations related to this gene include extracellular region; alpha-L-arabinofuranosidase activity; L-arabinose metabolic process; sensory perception of sound.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTOGL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTOGL
Gene Accession No. Q3ZCN5
Gene Alias C12orf64; DFNB84B; EG628870; Gm6924; otogelin like; otogelin-like; otogelin-like protein; otogelin-like, pseudogene 1; Otogl; Otogl-ps1; RGD1564294
Gene Symbols OTOGL
Host Species Rabbit
Immunogen Recombinant Human OTOGL. Recombinant protein control fragment (Product #RP-106461).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283310
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.