Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OTOP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144323
Description
OTOP1 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
Required for normal formation of otoconia in the inner ear. Inhibits P2Y purinoceptors. Modulates calcium homeostasis and influx of calcium in response to extracellular ATP.
Specifications
OTOP1 | |
Polyclonal | |
Unconjugated | |
OTOP1 | |
A530025J20Rik; hOtop1; OTOP1; otopetrin 1; otopetrin-1; Otp1; Proton channel OTOP1; tilted; tlt | |
Rabbit | |
Protein G | |
RUO | |
133060 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
2.98 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q7RTM1 | |
OTOP1 | |
Recombinant Human Proton channel OTOP1 protein (414-544AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction