Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTOP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144563 Supplier Invitrogen™ Supplier No. PA5144563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OTOP1 Polyclonal Antibody for Western Blot, ELISA

Required for normal formation of otoconia in the inner ear. Inhibits P2Y purinoceptors. Modulates calcium homeostasis and influx of calcium in response to extracellular ATP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTOP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.613 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene OTOP1
Gene Accession No. Q7ZWK8
Gene Alias A530025J20Rik; hOtop1; OTOP1; otopetrin 1; otopetrin-1; Otp1; Proton channel OTOP1; tilted; tlt
Gene Symbols OTOP1
Host Species Rabbit
Immunogen Recombinant Danio rerio (Zebrafish) (Brachydanio rerio) otop1 protein (406-518aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 322893
Target Species Zebrafish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.