Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTOP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548307
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548307 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548307 Supplier Invitrogen™ Supplier No. PA548307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with mouse based on sequence homology.

OTOP3 belongs to the otopetrin family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTOP3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OTOP3
Gene Accession No. Q7RTS5
Gene Alias 2310011E08Rik; Otop3; otopetrin 3; otopetrin-3; Proton channel OTOP3
Gene Symbols OTOP3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 536-564 amino acids from the C-terminal region of human OTOP3.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347741
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.