Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTP Polyclonal Antibody, Invitrogen™

Catalog No. PIPA584413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584413 Supplier Thermo Scientific Supplier No. PA584413

Rabbit Polyclonal Antibody

Immunogen sequence: GRQQAMAQSL SQCSLAAGPP PNSMGLSNSL AGSNGAGLQS HLYQPAFPGM VPASLPGPSN VSGSPQLCSS PDSSDVWRGT SIASLRRKAL EHTVSMSFT

OTP Polyclonal antibody specifically detects OTP in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTP
Gene Accession No. Q5XKR4
Gene Alias homeobox protein orthopedia; orthopedia homeobox; orthopedia homolog; orthopedia homolog (Drosophila); Otp
Gene Symbols OTP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OTP
Purification Method Anitgen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.