Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Otsuka America BreathTek™ UBT H. pylori Test Kit

Catalog No. 23655112 Shop All Meridian Bioscience Products
Click to view available options
Quantity:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Each kit includes one plastic tray with: Pranactin Citric drug powder (3g); self-adhesive bar code labels; blue bag (for baseline sample); pink bag (for post-dose sample); transport bag; plastic straw; drinking cup; instructions; and package insert.

No special training needed — CLIA nonregulated

For the qualitative detection of urease associated with H. pylori in the human stomach and as an aid in the initial diagnosis and post-treatment monitoring in adult patients.

  • Tests the entire gastric mucosa for active H. pylori infection, reducing the chance of false negatives
  • Sensitivity: 95%
  • Specificity: 95%
  • Non-invasive test — no blood or stool samples needed
  • Can be administered in office in 15 minutes; results in approximately 20 minutes
  • Four simple steps: patient exhales into blue bag, drinks the Pranactin™-Citric mixture, waits 15 minutes, exhales again into the pink bag

The Pranactin-Citric drug contains 84mg phenylalanine and is not recommended for patients with phenylketoneuria who cannot metabolize phenylalanine.

Specifications

Detectable Analytes Urease (H. pylori)
Type H. pylori Test Kit
Includes One Plastic tray with 3g Pranactin Citric drug powder, self-adhesive bar code labels, blue bag (for baseline sample), pink bag (for post-dose sample), transport bag, Plastic straw, drinking cup, instructions, package insert
Quantity Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.