Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OTUB1 Monoclonal Antibody (OTI1E3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526066
Description
OTUB1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The product of this gene is a member of the OTU superfamily of predicted cysteine proteases. The encoded protein is a highly specific ubiquitin iso-peptidase, and cleaves ubiquitin from branched poly-ubiquitin chains but not from ubiquitinated substrates. It interacts with another ubiquitin protease and an E3 ubiquitin ligase that inhibits cytokine gene transcription in the immune system. It is proposed to function in specific ubiquitin-dependent pathways, possibly by providing an editing function for polyubiquitin chain growth. Alternative splicing results in multiple transcript variants.
Specifications
OTUB1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96FW1 | |
OTUB1 | |
Full length human recombinant protein of OTUB1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI1E3 | |
Unconjugated | |
OTUB1 | |
AI850305; Deubiquitinating enzyme OTUB1; deubiquitinating enzyme OTUB1 {ECO:0000250; EMBL:AAI66771.1}; FLJ20113; FLJ40710; hOTU1; HSPC263; hypothetical protein LOC540984; MGC111158; MGC4584; OTB1; OTU deubiquitinase, ubiquitin aldehyde binding 1; OTU domain, ubiquitin aldehyde binding 1; OTU domain-containing ubiquitin aldehyde-binding protein 1; OTU domain-containing ubiquitin aldehyde-binding protein 1 {ECO:0000250; OTU1; OTUB1; otub1 {ECO:0000312; otubain 1; otubain-1; otubain-1 {ECO:0000250; OTU-domain Ubal-binding 1; Ubiquitin thioesterase OTUB1; ubiquitin thioesterase OTUB1 {ECO:0000250; ubiquitin-specific protease otubain 1; ubiquitin-specific-processing protease OTUB1; ubiquitin-specific-processing protease OTUB1 {ECO:0000250; UniProtKB:Q7TQI3} | |
Mouse | |
Affinity Chromatography | |
RUO | |
55611 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction