Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUB1 Monoclonal Antibody (OTI2H1)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526067

Catalog No. PIMA526067


Only null left
Add to Cart

Description

Description

OTUB1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The product of this gene is a member of the OTU superfamily of predicted cysteine proteases. The encoded protein is a highly specific ubiquitin iso-peptidase, and cleaves ubiquitin from branched poly-ubiquitin chains but not from ubiquitinated substrates. It interacts with another ubiquitin protease and an E3 ubiquitin ligase that inhibits cytokine gene transcription in the immune system. It is proposed to function in specific ubiquitin-dependent pathways, possibly by providing an editing function for polyubiquitin chain growth. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OTUB1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96FW1
OTUB1
Full length human recombinant protein of OTUB1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI2H1
Unconjugated
OTUB1
AI850305; Deubiquitinating enzyme OTUB1; deubiquitinating enzyme OTUB1 {ECO:0000250; EMBL:AAI66771.1}; FLJ20113; FLJ40710; hOTU1; HSPC263; hypothetical protein LOC540984; MGC111158; MGC4584; OTB1; OTU deubiquitinase, ubiquitin aldehyde binding 1; OTU domain, ubiquitin aldehyde binding 1; OTU domain-containing ubiquitin aldehyde-binding protein 1; OTU domain-containing ubiquitin aldehyde-binding protein 1 {ECO:0000250; OTU1; OTUB1; otub1 {ECO:0000312; otubain 1; otubain-1; otubain-1 {ECO:0000250; OTU-domain Ubal-binding 1; Ubiquitin thioesterase OTUB1; ubiquitin thioesterase OTUB1 {ECO:0000250; ubiquitin-specific protease otubain 1; ubiquitin-specific-processing protease OTUB1; ubiquitin-specific-processing protease OTUB1 {ECO:0000250; UniProtKB:Q7TQI3}
Mouse
Affinity Chromatography
RUO
55611
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.