Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUB1 Monoclonal Antibody (OTI5F7), TrueMAB™, OriGene
SDP

Catalog No. 502539851
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9851 30 μL
50-167-5730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9851 Supplier OriGene Supplier No. TA505204S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OTUB1 Monoclonal antibody specifically detects OTUB1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen OTUB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CDKN2B
Gene Accession No. P42772
Gene Alias AI850305; Deubiquitinating enzyme OTUB1; deubiquitinating enzyme OTUB1 {ECO:0000250; EMBL:AAI66771.1}; FLJ20113; FLJ40710; hOTU1; HSPC263; hypothetical protein LOC540984; MGC111158; MGC4584; OTB1; OTU deubiquitinase, ubiquitin aldehyde binding 1; OTU domain, ubiquitin aldehyde binding 1; OTU domain-containing ubiquitin aldehyde-binding protein 1; OTU domain-containing ubiquitin aldehyde-binding protein 1 {ECO:0000250; OTU1; OTUB1; otub1 {ECO:0000312; otubain 1; otubain-1; otubain-1 {ECO:0000250; OTU-domain Ubal-binding 1; Ubiquitin thioesterase OTUB1; ubiquitin thioesterase OTUB1 {ECO:0000250; ubiquitin-specific protease otubain 1; ubiquitin-specific-processing protease OTUB1; ubiquitin-specific-processing protease OTUB1 {ECO:0000250; UniProtKB:Q7TQI3}
Host Species Mouse
Immunogen Full length human recombinant protein of human CDKN2B produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1030
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.