Learn More
OTUB1 Mouse anti-Human, Clone: OTI1A6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505133

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The product of this gene is a member of the OTU superfamily of predicted cysteine proteases. The encoded protein is a highly specific ubiquitin iso-peptidase, and cleaves ubiquitin from branched poly-ubiquitin chains but not from ubiquitinated substrates. It interacts with another ubiquitin protease and an E3 ubiquitin ligase that inhibits cytokine gene transcription in the immune system. It is proposed to function in specific ubiquitin-dependent pathways, possibly by providing an editing function for polyubiquitin chain growth. Alternative splicing results in multiple transcript variants.Specifications
OTUB1 | |
Monoclonal | |
Unconjugated | |
OTUB1 | |
HSPC263, OTB1, OTU1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55611 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI1A6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96FW1 | |
OTUB1 | |
Full length human recombit protein of human OTUB1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.