Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTUB1 Mouse anti-Human, Clone: OTI1A6, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA600424

Description
The product of this gene is a member of the OTU superfamily of predicted cysteine proteases. The encoded protein is a highly specific ubiquitin iso-peptidase, and cleaves ubiquitin from branched poly-ubiquitin chains but not from ubiquitinated substrates. It interacts with another ubiquitin protease and an E3 ubiquitin ligase that inhibits cytokine gene transcription in the immune system. It is proposed to function in specific ubiquitin-dependent pathways, possibly by providing an editing function for polyubiquitin chain growth. Alternative splicing results in multiple transcript variants.Specifications
OTUB1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with <5% trehalose and 0.05% sodium azide | |
Q96FW1 | |
OTUB1 | |
Full length human recombit protein of human OTUB1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
OTI1A6 | |
Unconjugated | |
OTUB1 | |
HSPC263, OTB1, OTU1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55611 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction