Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578621 Supplier Thermo Scientific Supplier No. PA578621
Add to Cart
Add to Cart

OTUB1 Polyclonal Antibody

OTUB1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUB1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene OTUB1
Gene Accession No. B2RYG6, Q7TQI3
Gene Alias AI850305; Deubiquitinating enzyme OTUB1; deubiquitinating enzyme OTUB1 {ECO:0000250; EMBL:AAI66771.1}; FLJ20113; FLJ40710; hOTU1; HSPC263; hypothetical protein LOC540984; MGC111158; MGC4584; OTB1; OTU deubiquitinase, ubiquitin aldehyde binding 1; OTU domain, ubiquitin aldehyde binding 1; OTU domain-containing ubiquitin aldehyde-binding protein 1; OTU domain-containing ubiquitin aldehyde-binding protein 1 {ECO:0000250; OTU1; OTUB1; otub1 {ECO:0000312; otubain 1; otubain-1; otubain-1 {ECO:0000250; OTU-domain Ubal-binding 1; Ubiquitin thioesterase OTUB1; ubiquitin thioesterase OTUB1 {ECO:0000250; ubiquitin-specific protease otubain 1; ubiquitin-specific-processing protease OTUB1; ubiquitin-specific-processing protease OTUB1 {ECO:0000250; UniProtKB:Q7TQI3}
Gene Symbols OTUB1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of mouse OTUB1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107260, 293705
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.