Learn More
OTUB1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302918AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
The product of this gene is a member of the OTU superfamily of predicted cysteine proteases. The encoded protein is a highly specific ubiquitin iso-peptidase, and cleaves ubiquitin from branched poly-ubiquitin chains but not from ubiquitinated substrates. It interacts with another ubiquitin protease and an E3 ubiquitin ligase that inhibits cytokine gene transcription in the immune system. It is proposed to function in specific ubiquitin-dependent pathways, possibly by providing an editing function for polyubiquitin chain growth. Alternative splicing results in multiple transcript variants.Specifications
OTUB1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q7TQI3, Q96FW1 | |
OTUB1 | |
Between 40 and 80 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OTUB1 | |
OTUB1; OTU domain, ubiquitin aldehyde binding 1; FLJ20113; FLJ40710; HSPC263; MGC111158; MGC4584; OTB1; OTU1; OTU-domain Ubal-binding 1; otubain 1; ubiquitin-specific protease otubain 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
107260, 55611 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.