Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUB2 Monoclonal Antibody (OTI11C8), TrueMAB™, OriGene
SDP

Catalog No. 501673040
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-3040 100 μL
50-254-1900 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-3040 Supplier OriGene Supplier No. TA501945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hydrolase that can remove conjugated ubiquitin from proteins in vitro and may therefore play an important regulatory role at the level of protein turnover by preventing degradation. Mediates deubiquitination of ′Lys-11′-,′Lys-48′- and ′Lys-63′-linked polyubiquitin chains, with a preference for ′Lys-63′-linked polyubiquitin chains.

Specifications

Antigen OTUB2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI11C8
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene OTUB2
Gene Accession No. Q96DC9
Gene Alias C14orf137, OTB2, OTU2
Gene Symbols OTUB2
Host Species Mouse
Immunogen Full length human recombit protein of human OTUB2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78990
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.