Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OTUB2 Monoclonal Antibody (OTI2F6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501944S


Description
OTUB2 Monoclonal antibody specifically detects OTUB2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
OTUB2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96DC9 | |
Mouse | |
Affinity chromatography | |
RUO | |
78990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Western Blot | |
OTI2F6 | |
Unconjugated | |
OTUB2 | |
2010015L18Rik; 4930586I02Rik; AI413508; AW557219; C14orf137; deubiquitinating enzyme OTUB2; FLJ21916; MGC3102; OTB2; OTU deubiquitinase, ubiquitin aldehyde binding 2; OTU domain, ubiquitin aldehyde binding 2; OTU domain-containing ubiquitin aldehyde-binding protein 2; OTU2; OTUB2; Otubain-2; ubiquitin thioesterase OTUB2; ubiquitin-specific protease otubain 2; ubiquitin-specific-processing protease OTUB2 | |
Full length human recombinant protein of human OTUB2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction