Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUB2 Monoclonal Antibody (OTI6C7)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525303

Catalog No. PIMA525303


Only null left
Add to Cart

Description

Description

OTUB2 Monoclonal Antibody for Western Blot, Flow

Otubains are deubiquitylating cysteine proteases that belong to the ovarian tumor protein superfamily. Like other DUBs, otubains cleave proteins precisely at the ubiquitin -protein bond.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OTUB2
Monoclonal
0.56 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96DC9
OTUB2
Full length human recombinant protein of OTUB2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI6C7
Unconjugated
OTUB2
2010015L18Rik; 4930586I02Rik; AI413508; AW557219; C14orf137; deubiquitinating enzyme OTUB2; FLJ21916; MGC3102; OTB2; OTU deubiquitinase, ubiquitin aldehyde binding 2; OTU domain, ubiquitin aldehyde binding 2; OTU domain-containing ubiquitin aldehyde-binding protein 2; OTU2; OTUB2; Otubain-2; ubiquitin thioesterase OTUB2; ubiquitin-specific protease otubain 2; ubiquitin-specific-processing protease OTUB2
Mouse
Affinity Chromatography
RUO
78990
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.