Learn More
OTUB2 Mouse anti-Canine, Human, Rat, Clone: OTI11B3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501946

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Hydrolase that can remove conjugated ubiquitin from proteins in vitro and may therefore play an important regulatory role at the level of protein turnover by preventing degradation. Mediates deubiquitination of ′Lys-11′-,′Lys-48′- and ′Lys-63′-linked polyubiquitin chains, with a preference for ′Lys-63′-linked polyubiquitin chains.Specifications
OTUB2 | |
Monoclonal | |
Unconjugated | |
OTUB2 | |
C14orf137, OTB2, OTU2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
314405, 480420, 78990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI11B3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96DC9 | |
OTUB2 | |
Full length human recombit protein of human OTUB2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.