Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTUB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518961
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518961 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518961 Supplier Invitrogen™ Supplier No. PA518961
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

Otubains are deubiquitylating cysteine proteases that belong to the ovarian tumor protein superfamily. Like other DUBs, otubains cleave proteins precisely at the ubiquitin -protein bond.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUB2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene OTUB2
Gene Accession No. Q96DC9
Gene Alias 2010015L18Rik; 4930586I02Rik; AI413508; AW557219; C14orf137; deubiquitinating enzyme OTUB2; FLJ21916; MGC3102; OTB2; OTU deubiquitinase, ubiquitin aldehyde binding 2; OTU domain, ubiquitin aldehyde binding 2; OTU domain-containing ubiquitin aldehyde-binding protein 2; OTU2; OTUB2; Otubain-2; ubiquitin thioesterase OTUB2; ubiquitin-specific protease otubain 2; ubiquitin-specific-processing protease OTUB2
Gene Symbols OTUB2
Host Species Goat
Immunogen Synthetic peptide sequence (YKTSHYNILYAADKH) corresponding to the internal amino acids of OTUB2 (aa 220-234).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78990
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.