Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Otubain-2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300089F
Description
Otubain-2 Polyclonal specifically detects Otubain-2 in Human samples. It is validated for ELISA.Specifications
Otubain-2 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
78990 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
C14orf137chromosome 14 open reading frame 137, Deubiquitinating enzyme OTUB2, EC 3.4.19.12, FLJ21916, MGC3102, OTB2otubain-2, OTU domain, ubiquitin aldehyde binding 2, OTU domain-containing ubiquitin aldehyde-binding protein 2, OTU2ubiquitin thioesterase OTUB2, Otubain-2, ubiquitin-specific protease otubain 2, Ubiquitin-specific-processing protease OTUB2 | |
Produced in rabbits immunized with purified, recombinant Otubain-2 (Q96DC9-1; Met1-His234). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction